Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIV-1 gp120 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017774 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017774 0.1 mL
NB10066437 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017774 Supplier Novus Biologicals Supplier No. NB100664370.1ML
Add to Cart
Add to Cart

Goat Polyclonal Antibody

HIV-1 gp120 Polyclonal specifically detects HIV-1 gp120 in Virus samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot.

Specifications

Antigen HIV-1 gp120
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Formulation PBS (pH 7.2) with 0.09% Sodium Azide
Gene Accession No. Q70626
Gene Alias Envelope surface glycoprotein gp120, gp120, gp120 glycoprotein, Human immunodeficiency virus 1, SU, Surface protein
Gene Symbols env
Host Species Goat
Immunogen Purified gp120 from strain IIIB
Purification Method IgG purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 155971
Test Specificity NB100-66437 is specific for gp120, an exterior envelope glycoprotein on the surface of the virus particle that acts as a receptor for CD4 and chemokine receptors on macrophages and T-lymphocytes. It does not react with human T cells, B cells, normal bovine serum or normal human serum. In western blotting a predominant gp120 band is observed.
Target Species Virus
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.