Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIV-1 gp120 Antibody - BSA Free, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB10066437
Description
HIV-1 gp120 Polyclonal specifically detects HIV-1 gp120 in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
HIV-1 gp120 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.2) with 0.09% Sodium Azide | |
Envelope surface glycoprotein gp120, gp120, gp120 glycoprotein, Human immunodeficiency virus 1, SU, Surface protein | |
Goat | |
IgG purified | |
RUO | |
155971 | |
Virus | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen) | |
1.0 mg/mL | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
Q70626 | |
env | |
Purified gp120 from strain IIIB | |
0.5 mL | |
Primary | |
NB100-66437 is specific for gp120, an exterior envelope glycoprotein on the surface of the virus particle that acts as a receptor for CD4 and chemokine receptors on macrophages and T-lymphocytes. It does not react with human T cells, B cells, normal bovine serum or normal human serum. In western blotting a predominant gp120 band is observed. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction