Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIV-1 gp120 Antibody - BSA Free, Novus Biologicals™

Goat Polyclonal Antibody
$206.50 - $560.50
Specifications
Antigen | HIV-1 gp120 |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
Applications | Immunohistochemistry, ELISA, Immunohistochemistry (Frozen), Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017774
![]() |
Novus Biologicals
NB100664370.1ML |
0.1 mL |
Each for $206.50
|
|
|||||
NB10066437
![]() |
Novus Biologicals
NB10066437 |
0.5 mL |
Each for $560.50
|
|
|||||
Description
HIV-1 gp120 Polyclonal specifically detects HIV-1 gp120 in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
HIV-1 gp120 | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
Polyclonal | |
Purified | |
RUO | |
PBS (pH 7.2) with 0.09% Sodium Azide | |
Envelope surface glycoprotein gp120, gp120, gp120 glycoprotein, Human immunodeficiency virus 1, SU, Surface protein | |
env | |
IgG | |
IgG purified | |
NB100-66437 is specific for gp120, an exterior envelope glycoprotein on the surface of the virus particle that acts as a receptor for CD4 and chemokine receptors on macrophages and T-lymphocytes. It does not react with human T cells, B cells, normal bovine serum or normal human serum. In western blotting a predominant gp120 band is observed. |
1.0 mg/mL | |
Immunohistochemistry, ELISA, Immunohistochemistry (Frozen), Western Blot | |
Unconjugated | |
Goat | |
Virus | |
Q70626 | |
155971 | |
Purified gp120 from strain IIIB | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title