Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIV-1 gp120 Antibody, DyLight 755, Novus Biologicals™
SDP

Catalog No. NB206753 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB206753 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB206753 Supplier Novus Biologicals Supplier No. NBP176371IR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HIV-1 gp120 Polyclonal antibody specifically detects HIV-1 gp120 in Virus samples. It is validated for Western Blot, ELISA

Specifications

Antigen HIV-1 gp120
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias Envelope surface glycoprotein gp120, gp120, gp120 glycoprotein, Human immunodeficiency virus 1, SU, Surface protein
Host Species Rabbit
Immunogen Antibody was raised against 16 amino acid synthetic peptide representing the major HIV-1 consensus sequence near the N-terminal region of V1/V2 loop. The immunogen is located within amino acids 80 - 130 of gp120. Amino Acid Squence: EQMHEDIISLWDQLSK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 155971
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.