Learn More
HIV-1 gp41 Antibody (402D9.14/clone 1), Alexa Fluor™ 546, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | HIV-1 gp41 |
| Applications | ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | 402D9.14/clone 1 |
| Conjugate | Alexa Fluor 546 |
| Dilution | Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 |
| Gene Alias | Env, Glycoprotein 41, gp41, Human immunodeficiency virus 1, Human Immunodeficiency Virus Type 1 gp41, TM, Transmembrane protein |
| Host Species | Mouse |
| Immunogen | HR1-PID-HR2-transfected HEK 293 cells (HIV-1, 92BR025, Clade C) |
| Purification Method | Ion exchange chromatography |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.