Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIV-1 gp41 Antibody (402D9.14/clone 1), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. DDX1304A647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DDX1304A647 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DDX1304A647 Supplier Novus Biologicals Supplier No. DDX1304A647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HIV-1 gp41 Monoclonal specifically detects HIV-1 gp41 in Virus samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen HIV-1 gp41
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 402D9.14/clone 1
Conjugate Alexa Fluor 647
Dilution Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000
Gene Alias Env, Glycoprotein 41, gp41, Human immunodeficiency virus 1, Human Immunodeficiency Virus Type 1 gp41, TM, Transmembrane protein
Host Species Mouse
Immunogen HR1-PID-HR2-transfected HEK 293 cells (HIV-1, 92BR025, Clade C)
Purification Method Ion exchange chromatography
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at -20C in the dark. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.