Learn More
Description
Specifications
Specifications
| Antigen | HIV-1 gp41 |
| Applications | ELISA, Flow Cytometry |
| Classification | Monoclonal |
| Clone | 425D11.01/ clone 3 |
| Conjugate | Biotin |
| Dilution | Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 |
| Gene Alias | Env, Glycoprotein 41, gp41, Human immunodeficiency virus 1, Human Immunodeficiency Virus Type 1 gp41, TM, Transmembrane protein |
| Host Species | Mouse |
| Immunogen | HR1-PID-HR2-transfected HEK 293 cells (HIV-1, 92BR025, Clade C) |
| Purification Method | Ion exchange chromatography |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
