Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIV-1 Protease Antibody (1696), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB500337R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500337R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500337R Supplier Novus Biologicals Supplier No. NB500337R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HIV-1 Protease Monoclonal antibody specifically detects HIV-1 Protease in Virus samples. It is validated for Western Blot, ELISA.

Specifications

Antigen HIV-1 Protease
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1696
Conjugate DyLight 550
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Host Species Mouse
Immunogen Bacterially expressed full-length HIV-1 protease
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity The antibody 1696 recognizes free N-terminus of mature HIV protease (HIV-1 and HIV-2), an enzyme that hydrolyzes polyproteins of HIV viruses into functional proteins. The antibody 1696 does not react with the precursor.
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.