Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIV-1 Rev binding protein Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Applications | Western Blot |
|---|---|
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Regulatory Status | RUO |
Description
HIV-1 Rev binding protein Polyclonal specifically detects HIV-1 Rev binding protein in Human samples. It is validated for Western Blot.Specifications
| Western Blot | |
| Unconjugated | |
| RUO | |
| ArfGAP with FG repeats 1, DKFZp686I15205, HIV-1 Rev-binding protein, HRBRev interacting protein, Rab, Rev/Rex activation domain-binding protein, RABMGC116938, RIPMGC116940 | |
| AGFG1 | |
| Primary |
| Polyclonal | |
| Rabbit | |
| P52594 | |
| 3267 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title