Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIV-1 Tat specific factor 1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP106041 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP106041 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP106041 Supplier Novus Biologicals Supplier No. NBP106041
Add to Cart
Add to Cart

Goat Polyclonal Antibody

HIV-1 Tat specific factor 1 Polyclonal antibody specifically detects HIV-1 Tat specific factor 1 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen HIV-1 Tat specific factor 1
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:16000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_055315.2
Gene Alias cofactor required for Tat activation of HIV-1 transcription, dJ196E23.2, HIV TAT specific factor 1, HIV Tat-specific factor 1, HIV-1 Tat specific factor 1, TAT-SF1
Gene Symbols HTATSF1
Host Species Goat
Immunogen Peptide with sequence C-QELYGDGKDGDTQTD corresponding to internal region of HIV-1 Tat specific factor 1 (near N-Terminus) according to NP_055315.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27336
Test Specificity Cross reacts with Human. Expected from sequence similarity: Rat and Dog.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.