Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIVEP2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP146125 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP146125 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP146125 Supplier Novus Biologicals Supplier No. NBP146125
Add to Cart
Add to Cart

Goat Polyclonal Antibody

HIVEP2 Polyclonal specifically detects HIVEP2 in Human, Mouse, Rat, Bovine, Canine samples. It is validated for Peptide ELISA.

Specifications

Antigen HIVEP2
Applications ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Peptide ELISA Detection limit 1:1000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_006725.3
Gene Alias c-myc intron binding protein 1, c-myc intron-binding protein 1, human immunodeficiency virus type I enhancer binding protein 2, Human immunodeficiency virus type I enhancer-binding protein 2HIV-EP2MBP-2SHN2, MHC binding protein-2, MHC-binding protein 2, MIBP1, Schnurri-2, transcription factor HIVEP2, ZAS2, ZNF40B
Gene Symbols HIVEP2
Host Species Goat
Immunogen Peptide with sequence C-TEESKDPSSEKSQ corresponding to C-Terminus according to NP_006725.3.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3097
Test Specificity This antibody is specific to the C-Terminus.
Target Species Human, Mouse, Rat, Bovine, Canine
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.