Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HJURP Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB167035 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167035 100 μg
NB167036 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB167035 Supplier Novus Biologicals Supplier No. NBP315970100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HJURP Polyclonal antibody specifically detects HJURP in Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen HJURP
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000 - 1:2000
Formulation PBS, 50% glycerol, pH7.3
Gene Alias FAKTS14-3-3-associated AKT substrate, fetal liver expressing gene 1, Fetal liver-expressing gene 1 protein, FLEG1, hFLEG1, Holliday junction recognition protein, Up-regulated in lung cancer 9, URLC9DKFZp762E1312
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-190 of human HJURP (NP_060880.3). MLGTLRAMEGEDVEDDQLLQKLRASRRRFQRRMQRLIEKYNQPFEDTPVVQMATLTYETPQGLRIWGGRLIKERNEGEIQDSSMKPADRTDGSVQAAAWGPELPSHRTVLGADSKSGEVDATSDQEESVAWALAPAVPQSPLKNELRRKYLTQVDILLQGAEYFECAGNRAGRDVRVTPLPSLASPAVPA
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55355
Target Species Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.