Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HJURP Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$148.20 - $378.00
Specifications
Antigen | HJURP |
---|---|
Dilution | Western Blot 1:1000 - 1:2000 |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
HJURP Polyclonal antibody specifically detects HJURP in Mouse, Rat samples. It is validated for Western BlotSpecifications
HJURP | |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Rat | |
FAKTS14-3-3-associated AKT substrate, fetal liver expressing gene 1, Fetal liver-expressing gene 1 protein, FLEG1, hFLEG1, Holliday junction recognition protein, Up-regulated in lung cancer 9, URLC9DKFZp762E1312 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-190 of human HJURP (NP_060880.3). MLGTLRAMEGEDVEDDQLLQKLRASRRRFQRRMQRLIEKYNQPFEDTPVVQMATLTYETPQGLRIWGGRLIKERNEGEIQDSSMKPADRTDGSVQAAAWGPELPSHRTVLGADSKSGEVDATSDQEESVAWALAPAVPQSPLKNELRRKYLTQVDILLQGAEYFECAGNRAGRDVRVTPLPSLASPAVPA | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot 1:1000 - 1:2000 | |
Polyclonal | |
Purified | |
RUO | |
PBS, 50% glycerol, pH7.3 | |
55355 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title