Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HKR2 Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB146620 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB146620 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB146620 Supplier Novus Biologicals Supplier No. NBP306056AF532
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HKR2 Polyclonal specifically detects HKR2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen HKR2
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias GLI-Kruppel family member HKR2, HKR2MGC138482, Krueppel-related zinc finger protein 2, Protein HKR2, zinc finger and SCAN domain containing 22, zinc finger and SCAN domain-containing protein 22, Zinc finger protein 50MGC126679, ZNF50
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human HKR2 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 342945
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.