Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA A Antibody (108-2C5) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB121507 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB121507 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB121507 Supplier Novus Biologicals Supplier No. NBP311420
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HLA A Monoclonal antibody specifically detects HLA A in Human samples. It is validated for Flow Cytometry, Immunofluorescence, Immunofluorescence

Specifications

Antigen HLA A
Applications Flow Cytometry, Immunofluorescence, Immunofluorescence
Classification Monoclonal
Clone 108-2C5
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml
Formulation 10 mM PBS
Gene Alias A-10 alpha chain, FLJ26655, HLA class I histocompatibility antigen, A-1 alpha chain, HLA class I histocompatibility antigen, A-28 alpha chain, HLA class I histocompatibility antigen, A-9 alpha chain, HLAA, major histocompatibility complex, class I, A, MHC class I antigen A*1, MHC class I antigen A*11, MHC class I antigen A*80
Host Species Mouse
Immunogen Normal human peripheral blood lymphocytes
Purification Method Bioreactor concentrate
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cell Biology, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3105
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.