Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA A Antibody (SR1691), Novus Biologicals™
SDP

Catalog No. NB306091 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB306091 50 μL
NB306092 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB306091 Supplier Novus Biologicals Supplier No. NBP32164250UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

HLA A Monoclonal antibody specifically detects HLA A in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation

Specifications

Antigen HLA A
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone SR1691
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias A-10 alpha chain, FLJ26655, HLA class I histocompatibility antigen, A-1 alpha chain, HLA class I histocompatibility antigen, A-28 alpha chain, HLA class I histocompatibility antigen, A-9 alpha chain, HLAA, major histocompatibility complex, class I, A, MHC class I antigen A*1, MHC class I antigen A*11, MHC class I antigen A*80
Host Species Rabbit
Immunogen A synthesized peptide derived from human HLA A (Uniprot #: P01891)
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cell Biology, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3105
Target Species Human
Content And Storage Store at -20 degrees C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.