Learn More
Description
Specifications
Specifications
| Antigen | HLA A2 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | BB7.2 |
| Conjugate | Alexa Fluor 488 |
| Dilution | Flow Cytometry |
| Gene Alias | A-10 alpha chain, FLJ26655, HLA class I histocompatibility antigen, A-1 alpha chain, HLA class I histocompatibility antigen, A-28 alpha chain, HLA class I histocompatibility antigen, A-9 alpha chain, HLAA, major histocompatibility complex, class I, A, MHC class I antigen A*1, MHC class I antigen A*11, MHC class I antigen A*80 |
| Gene Symbols | HLA-A |
| Host Species | Mouse |
| Immunogen | HLA-A2 solubilised by papain |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
