Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA A2 Antibody (BB7.2), PE/Cy7, Novus Biologicals™
SDP

Catalog No. N144896PEC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N144896PEC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N144896PEC7 Supplier Novus Biologicals Supplier No. NBP144896PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HLA A2 Monoclonal specifically detects HLA A2 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen HLA A2
Applications Flow Cytometry
Classification Monoclonal
Clone BB7.2
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias A-10 alpha chain, FLJ26655, HLA class I histocompatibility antigen, A-1 alpha chain, HLA class I histocompatibility antigen, A-28 alpha chain, HLA class I histocompatibility antigen, A-9 alpha chain, HLAA, major histocompatibility complex, class I, A, MHC class I antigen A*1, MHC class I antigen A*11, MHC class I antigen A*80
Gene Symbols HLA-A
Host Species Mouse
Immunogen HLA-A2 solubilised by papain
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Adaptive Immunity, Cell Biology, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3105.0
Test Specificity The antibody BB7.2 recognizes an epitope at the C-terminus of alpha-2 helix and a turn on one of the underlying beta strands within the human HLA-A2 histocompatibility antigen.
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.