Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA ABC Antibody (W6/32), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB10064775G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064775G 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10064775G Supplier Novus Biologicals Supplier No. NB10064775G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HLA ABC Monoclonal specifically detects HLA ABC in Human samples. It is validated for CyTOF, ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) (Negative), Immunoprecipitation.

Specifications

Antigen HLA ABC
Applications CyTOF, ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) (Negative), Immunoprecipitation
Classification Monoclonal
Clone W6/32
Conjugate DyLight 488
Dilution Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Gene Alias A-10 alpha chain, FLJ26655, HLA class I histocompatibility antigen, A-1 alpha chain, HLA class I histocompatibility antigen, A-28 alpha chain, HLA class I histocompatibility antigen, A-9 alpha chain, HLAA, major histocompatibility complex, class I, A, MHC class I antigen A*1, MHC class I antigen A*11, MHC class I antigen A*80
Gene Symbols HLA-A
Host Species Mouse
Immunogen Purified human tonsil lymphocyte membranes.
Purification Method Protein G purified
Quantity 0.25 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cell Biology, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3105
Test Specificity NB100-64775 recognizes an antigenic determinant shared among products of the HLA A, B and C loci. Clone W6/32 recognizes a conformational epitope, reacting with HLA class I alpha3 and alpha2 domains (6). MHC class I glycoproteins are found on the surface of almost all nucleated somatic cells. This product is routinely tested in flow cytometry on human peripheral blood lymphocytes.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.