Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA ABC Antibody (YTH862.2), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB065411549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB065411549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB065411549 Supplier Novus Biologicals Supplier No. NB10065411JF549
Add to Cart
Add to Cart

Rat Monoclonal Antibody

HLAABC Monoclonal specifically detects HLAABC in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation.

Specifications

Antigen HLAABC
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone YTH862.2
Conjugate Janelia Fluor 549
Dilution Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias A-10 alpha chain, FLJ26655, HLA class I histocompatibility antigen, A-1 alpha chain, HLA class I histocompatibility antigen, A-28 alpha chain, HLA class I histocompatibility antigen, A-9 alpha chain, HLAA, major histocompatibility complex, class I, A, MHC class I antigen A*1, MHC class I antigen A*11, MHC class I antigen A*80
Gene Symbols HLA-A
Host Species Rat
Immunogen PHA activated human peripheral blood lymphocytes.
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Adaptive Immunity, Cell Biology, Immunology, Major Histocompatibility Complexes
Primary or Secondary Primary
Gene ID (Entrez) 3105
Test Specificity NB100-65411 recognizes a monomorphic determinant on the human HLA Class I heavy chain. Epitope analysis has shown that this antibody binds in the alpha1 domain. MHC Class I is expressed by all nucleated cells. Recent studies have shown that this antibody may be able to induce apoptosis in activated but not resting T cells (1).
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.