Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA-ABCE Antibody (TP25.99SF), PE/Atto594, Novus Biologicals™
SDP

Catalog No. N26806PE594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N26806PE594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N26806PE594 Supplier Novus Biologicals Supplier No. NBP268006PEATT594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HLA-ABCE Monoclonal specifically detects HLA-ABCE in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen HLA-ABCE
Applications Flow Cytometry
Classification Monoclonal
Clone TP25.99SF
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias HLA-ABC, HLA-E, major histocompatibility complex, class I, A, major histocompatibility complex, class I, B, major histocompatibility complex, class I, C, major histocompatibility complex, class I, E, MHC class I
Host Species Mouse
Immunogen IFN-gamma-treated human melanoma cells COLO 38.
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity The mouse monoclonal antibody TP25.99SF recognizes HLA-ABC and HLA-E molecules, but not HLA-G. It can be used for distinguishing between HLA-G and other HLA-class I antigens.
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.