Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA-B Rat anti-Human, Alexa Fluor™ 647, Clone: YTH 76.3.rMAb (also known as YTH/76.3; YT76), BD Phrmingen™
Rat Monoclonal Antibody
$110.50 - $273.50
Specifications
Antigen | HLA-B |
---|---|
Clone | YTH 76.3.rMAb (also known as YTH/76.3; YT76) |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Alexa Fluor 647 |
Description
HLA-B Monoclonal antibody specifically detects HLA-B in Human samples. It is validated for Flow cytometrySpecifications
HLA-B | |
Flow Cytometry | |
Alexa Fluor 647 | |
Rat | |
Human | |
HLA-B; HLAB | |
Normal Human Lymphocytes | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. |
YTH 76.3.rMAb (also known as YTH/76.3; YT76) | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
3106 | |
IgG1 κ | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title