Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA B7 Antibody (BB7.1), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB10064159I Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064159I 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10064159I Supplier Novus Biologicals Supplier No. NB10064159IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HLA B7 Monoclonal specifically detects HLA B7 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin).

Specifications

Antigen HLA B7
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone BB7.1
Conjugate DyLight 755
Dilution Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
Gene Accession No. P01889
Gene Alias ankylosing spondylitis, AS, b'DT, Bw-22, Bw-4, Bw-41, Bw-44, Bw-45, Bw-46, Bw-47, Bw-48, Bw-50, Bw-52, Bw-53, Bw-54, Bw-55, Bw-56, Bw-57, Bw-58, Bw-60, HLA class I histocompatibility antigen, B alpha chain, HLA class I histocompatibility antigen, B-12 alpha chain, HLA class I histocompatibility antigen, B-21 alpha chain, HLA class I histocompatibility antigen, B-5 alpha chain, HLA class I histocompatibility antigen, B-7 alpha chain, HLAB, HLA-B27, HLA-B73, leukocyte antigen class I-B, lymphocyte antigen, major histocompatibility complex, class I, B, MGC111087, MHC class I antigen B*13, MHC class I antigen B*14, MHC class I antigen B*15, MHC class I antigen B*18, MHC class I antigen B*27, MHC class I antigen B*35, MHC class I antigen B*37, MHC class I antigen B*38, MHC class I antigen B*39, MHC class I antigen B*40, MHC class I antigen B*41, MHC class I antigen B*42, MHC class I antigen B*44, MHC class I antigen B*45, MHC class I antigen B*46, MHC class I antigen B*47, MHC class I anti
Gene Symbols HLA-B
Host Species Mouse
Immunogen Papain solubilized HLA-B7 antigen
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3106
Test Specificity Recognizes the HLA-B7 antigen and does not cross-react with HLA-B27 or other related antigens. It can be used to distinguish true HLA-B27 positives from false HLA-B27 positives (i.e. HLA-B7 positive) in the investigation of diseases such as ankylosing spondylitis and anterior uveitis.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.