Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA B7 Rabbit anti-Human, DyLight 650, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB132503 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB132503 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB132503 Supplier Novus Biologicals Supplier No. NBP306118C

Rabbit Polyclonal Antibody

HLA B7 Polyclonal specifically detects HLA B7 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen HLA B7
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias ankylosing spondylitis, AS, b'DT, Bw-22, Bw-4, Bw-41, Bw-44, Bw-45, Bw-46, Bw-47, Bw-48, Bw-50, Bw-52, Bw-53, Bw-54, Bw-55, Bw-56, Bw-57, Bw-58, Bw-60, HLA class I histocompatibility antigen, B alpha chain, HLA class I histocompatibility antigen, B-12 alpha chain, HLA class I histocompatibility antigen, B-21 alpha chain, HLA class I histocompatibility antigen, B-5 alpha chain, HLA class I histocompatibility antigen, B-7 alpha chain, HLAB, HLA-B27, HLA-B73, leukocyte antigen class I-B, lymphocyte antigen, major histocompatibility complex, class I, B, MGC111087, MHC class I antigen B*13, MHC class I antigen B*14, MHC class I antigen B*15, MHC class I antigen B*18, MHC class I antigen B*27, MHC class I antigen B*35, MHC class I antigen B*37, MHC class I antigen B*38, MHC class I antigen B*39, MHC class I antigen B*40, MHC class I antigen B*41, MHC class I antigen B*42, MHC class I antigen B*44, MHC class I antigen B*45, MHC class I antigen B*46, MHC class I antigen B*47, MHC class I anti
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human HLA B7.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3106
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.