Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA C Antibody (DT-9), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP20419A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP20419A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP20419A30 Supplier Novus Biologicals Supplier No. NBP250419AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HLA C Monoclonal antibody specifically detects HLA C in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen HLA C
Applications Flow Cytometry
Classification Monoclonal
Clone DT-9
Conjugate Alexa Fluor 350
Dilution Flow Cytometry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias C alpha chain, D6S204, FLJ27082, HLA class I histocompatibility antigen, Cw-1 alpha chain, HLAC, HLA-Cw, HLA-JY3, HLC-C, human leukocyte antigen-C alpha chain, leucocyte antigen C, leucocyte antigen Cw, leukocyte antigen-C alpha chain, major histocompatibility antigen HLA-C, major histocompatibility complex, class I, C, MHC class I antigen Cw*1, MHC class I antigen Cw*12, MHC class I antigen Cw*14, MHC class I antigen Cw*15, MHC class I antigen Cw*16, MHC class I antigen Cw*17, MHC class I antigen Cw*2, MHC class I antigen Cw*3, MHC class I antigen Cw*4, MHC class I antigen Cw*5, MHC class I antigen Cw*6, MHC class I antigen Cw*7, MHC class I antigen Cw*8, MHC class I antigen heavy chain HLA-C, PSORS1
Gene Symbols HLA-C
Host Species Mouse
Immunogen Purified tamarin MHC class I molecules (using anti-class I antibody BB7.7)
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3107
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.