Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA C Antibody (DT-9), FITC, Novus Biologicals™
SDP

Catalog No. NB305670 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB305670 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB305670 Supplier Novus Biologicals Supplier No. FAB10394F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HLA C Monoclonal antibody specifically detects HLA C in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen HLA C
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone DT-9
Conjugate FITC
Dilution Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias C alpha chain, D6S204, FLJ27082, HLA class I histocompatibility antigen, Cw-1 alpha chain, HLAC, HLA-Cw, HLA-JY3, HLC-C, human leukocyte antigen-C alpha chain, leucocyte antigen C, leucocyte antigen Cw, leukocyte antigen-C alpha chain, major histocompatibility antigen HLA-C, major histocompatibility complex, class I, C, MHC class I antigen Cw*1, MHC class I antigen Cw*12, MHC class I antigen Cw*14, MHC class I antigen Cw*15, MHC class I antigen Cw*16, MHC class I antigen Cw*17, MHC class I antigen Cw*2, MHC class I antigen Cw*3, MHC class I antigen Cw*4, MHC class I antigen Cw*5, MHC class I antigen Cw*6, MHC class I antigen Cw*7, MHC class I antigen Cw*8, MHC class I antigen heavy chain HLA-C, PSORS1
Host Species Mouse
Immunogen Purified tamarin MHC class I molecules (using anti-class I antibody BB7.7), Accession # P10321
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3107
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.