Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA Class I Antibody (MEM-123), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB50505A750 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB50505A750 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB50505A750 Supplier Novus Biologicals Supplier No. NB500505AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HLA Class I Monoclonal specifically detects HLA Class I in Human, Bovine, Primate samples. It is validated for Flow Cytometry, ELISA.

Specifications

Antigen HLA Class I
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone MEM-123
Conjugate Alexa Fluor 750
Dilution Flow Cytometry, ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias A-10 alpha chain, FLJ26655, HLA class I histocompatibility antigen, A-1 alpha chain, HLA class I histocompatibility antigen, A-28 alpha chain, HLA class I histocompatibility antigen, A-9 alpha chain, HLAA, major histocompatibility complex, class I, A, MHC class I antigen A*1, MHC class I antigen A*11, MHC class I antigen A*80
Gene Symbols HLA-A
Host Species Mouse
Immunogen COS-7 African green monkey kidney cells
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3105
Test Specificity This antibody (clone MEM-123) reacts with all human classical MHC Class I molecules (major histocompatibility complex) in native cell-surface forms as well as with human HLA-G cDNA transfected cells. MHC Class I molecules (MHC Class Ia) are expressed on the surface of all human cell types. This antibody completely blocks binding of classical W6/32 to surface-expressed HLA-G, but does not cross-blocks This antibody MEM-G/9.
Target Species Human, Bovine, Primate
Content And Storage Store at 4°C in the dark.
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.