Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA Class I Antibody (MEM-123), FITC, Novus Biologicals™
SDP

Catalog No. NB359192 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB359192 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB359192 Supplier Novus Biologicals Supplier No. NB500505F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HLA Class I Monoclonal antibody specifically detects HLA Class I in Human, Bovine, Primate samples. It is validated for Flow Cytometry, ELISA, Immunoprecipitation, CyTOF

Specifications

Antigen HLA Class I
Applications Flow Cytometry, ELISA, Immunoprecipitation, CyTOF
Classification Monoclonal
Clone MEM-123
Conjugate FITC
Dilution Flow Cytometry, ELISA, Immunoprecipitation, CyTOF-ready
Formulation PBS
Gene Alias A-10 alpha chain, FLJ26655, HLA class I histocompatibility antigen, A-1 alpha chain, HLA class I histocompatibility antigen, A-28 alpha chain, HLA class I histocompatibility antigen, A-9 alpha chain, HLAA, major histocompatibility complex, class I, A, MHC class I antigen A*1, MHC class I antigen A*11, MHC class I antigen A*80
Host Species Mouse
Immunogen COS-7 African green monkey kidney cells
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cell Biology, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3105
Target Species Human, Bovine, Primate
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.