Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA Class I Antibody (MEM-147), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB50411A750 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB50411A750 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB50411A750 Supplier Novus Biologicals Supplier No. NB500411AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HLA Class I Monoclonal specifically detects HLA Class I in Human samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen HLA Class I
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone MEM-147
Conjugate Alexa Fluor 750
Dilution Western Blot, Flow Cytometry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias A-10 alpha chain, FLJ26655, HLA class I histocompatibility antigen, A-1 alpha chain, HLA class I histocompatibility antigen, A-28 alpha chain, HLA class I histocompatibility antigen, A-9 alpha chain, HLAA, major histocompatibility complex, class I, A, MHC class I antigen A*1, MHC class I antigen A*11, MHC class I antigen A*80
Gene Symbols HLA-A
Host Species Mouse
Immunogen PHA-activated peripheral blood lymphocytes
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3105
Test Specificity This antibody (clone MEM-147) reacts with all human classical MHC Class I molecules in native cell-surface forms (e.g. it recognizes native HLA-A2 in cytofluorometry and immunoprecipitation but not in Western blotting). MHC Class I molecules (MHC Class Ia) are expressed on the surface of all human nucleated cell types. The antibody MEM-147 is positive in Western blotting (non-reducing conditions) only with most HLA-B and HLA-C molecules, but not HLA-A. Reactivity is very similar to the classical antibody W6/32.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.