Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA Class I Antibody (MEM-81) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB500412 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500412 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500412 Supplier Novus Biologicals Supplier No. NB500412
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HLA Class I Monoclonal specifically detects HLA Class I in Human samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen HLA Class I
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone MEM-81
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1-5 ug/ml, CyTOF-ready
Gene Accession No. P04439
Gene Alias A-10 alpha chain, FLJ26655, HLA class I histocompatibility antigen, A-1 alpha chain, HLA class I histocompatibility antigen, A-28 alpha chain, HLA class I histocompatibility antigen, A-9 alpha chain, HLAA, major histocompatibility complex, class I, A, MHC class I antigen A*1, MHC class I antigen A*11, MHC class I antigen A*80
Gene Symbols HLA-A
Host Species Mouse
Immunogen Leukocytes of a patient suffering with LGL type leukemia.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cell Biology, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3105
Test Specificity The antibody MEM-81 reacts with human classical MHC Class I molecules in native cell-surface forms. MHC Class I molecules (MHC Class Ia) are expressed on the surface of human nucleated cell types.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.