Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA Class I Antibody (W6/32), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB302827 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB302827 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB302827 Supplier Novus Biologicals Supplier No. FAB7098MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HLA Class I Monoclonal antibody specifically detects HLA Class I in Human samples. It is validated for Flow Cytometry, CyTOF-reported

Specifications

Antigen HLA Class I
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone W6/32
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry, CyTOF-reported
Formulation 50mM Sodium Borate
Gene Alias A-10 alpha chain, FLJ26655, HLA class I histocompatibility antigen, A-1 alpha chain, HLA class I histocompatibility antigen, A-28 alpha chain, HLA class I histocompatibility antigen, A-9 alpha chain, HLAA, major histocompatibility complex, class I, A, MHC class I antigen A*1, MHC class I antigen A*11, MHC class I antigen A*80
Host Species Mouse
Immunogen Membranes from human tonsillar lymphocytes,
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cell Biology, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3105
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.