Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA DMA Antibody (5C9NB), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP24430A594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP24430A594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP24430A594 Supplier Novus Biologicals Supplier No. NBP244300AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HLA DMA Monoclonal specifically detects HLA DMA in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Cell Surface).

Specifications

Antigen HLA DMA
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 5C9NB
Conjugate Alexa Fluor 594
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Cell Surface)
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias class II histocompatibility antigen, M alpha chain, D6S222E, DMA, HLA class II histocompatibility antigen, DM alpha chain, major histocompatibility complex, class II, DM alpha, MHC class II antigen DMA, Really interesting new gene 6 protein, RING6HLADM
Gene Symbols HLA-DMA
Host Species Mouse
Immunogen Partial recombinant human HLA DMA protein (between amino acids 10-300) [Entrez NP_006111]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3108
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.