Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA DMA Antibody (6B12NB) - Azide Free, Novus Biologicals™
SDP

Catalog No. NBP244304 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP244304 0.1 mg
NBP244304SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP244304 Supplier Novus Biologicals Supplier No. NBP244304
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ensure accurate, reproducible results in Western Blot, Flow (Cell Surface), Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)

HLA DMA Monoclonal specifically detects HLA DMA in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Cell Surface).

Specifications

Antigen HLA DMA
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 6B12NB
Concentration 1.0mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 ug/ml, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 10-20 ug/ml, Immunohistochemistry-Paraffin 1:200, Flow (Cell Surface) 2.5 ug/million cells
Formulation PBS with No Preservative
Gene Alias class II histocompatibility antigen, M alpha chain, D6S222E, DMA, HLA class II histocompatibility antigen, DM alpha chain, major histocompatibility complex, class II, DM alpha, MHC class II antigen DMA, Really interesting new gene 6 protein, RING6HLADM
Gene Symbols HLA-DMA
Host Species Mouse
Immunogen Partial recombinant human HLA DMA protein (between amino acids 10-300) [Entrez NP_006111]
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Asthma, Diabetes Research, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3108
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.