Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA DMA Antibody (6B12NB) - Azide Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $499.50
Specifications
Antigen | HLA DMA |
---|---|
Clone | 6B12NB |
Concentration | 1.0mg/mL |
Dilution | Western Blot 2 ug/ml, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 10-20 ug/ml, Immunohistochemistry-Paraffin 1:200, Flow (Cell Surface) 2.5 ug/million cells |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP244304SS
![]() |
Novus Biologicals
NBP244304SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP244304
![]() |
Novus Biologicals
NBP244304 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
Ensure accurate, reproducible results in Western Blot, Flow (Cell Surface), Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
HLA DMA Monoclonal specifically detects HLA DMA in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Cell Surface).Specifications
HLA DMA | |
1.0mg/mL | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Asthma, Diabetes Research, Immunology | |
PBS with No Preservative | |
3108 | |
Partial recombinant human HLA DMA protein (between amino acids 10-300) [Entrez NP_006111] | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
6B12NB | |
Western Blot 2 ug/ml, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 10-20 ug/ml, Immunohistochemistry-Paraffin 1:200, Flow (Cell Surface) 2.5 ug/million cells | |
Monoclonal | |
Purified | |
RUO | |
Human | |
class II histocompatibility antigen, M alpha chain, D6S222E, DMA, HLA class II histocompatibility antigen, DM alpha chain, major histocompatibility complex, class II, DM alpha, MHC class II antigen DMA, Really interesting new gene 6 protein, RING6HLADM | |
HLA-DMA | |
IgG1 κ | |
Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title