Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA DMA Antibody (6B12NB), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB336971 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB336971 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB336971 Supplier Novus Biologicals Supplier No. NBP244302MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HLA DMA Monoclonal antibody specifically detects HLA DMA in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Flow (Cell Surface)

Specifications

Antigen HLA DMA
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Flow Cytometry (Cell Surface)
Classification Monoclonal
Clone 6B12NB
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Cell Surface)
Formulation 50mM Sodium Borate
Gene Alias class II histocompatibility antigen, M alpha chain, D6S222E, DMA, HLA class II histocompatibility antigen, DM alpha chain, major histocompatibility complex, class II, DM alpha, MHC class II antigen DMA, Really interesting new gene 6 protein, RING6HLADM
Host Species Mouse
Immunogen Partial recombinant human HLA DMA protein (between amino acids 10-300) [Entrez NP_006111]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Asthma, Diabetes Research, Immunology, Lipid and Metabolism, Membrane Trafficking and Chaperones, Neuroendocrinology
Primary or Secondary Primary
Gene ID (Entrez) 3108
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.