Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA DMB Antibody, Novus Biologicals™
SDP

Catalog No. NB406652 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB406652 25 μL
NBP185326 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB406652 Supplier Novus Biologicals Supplier No. NBP18532625UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HLA DMB Polyclonal specifically detects HLA DMB in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen HLA DMB
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias class II histocompatibility antigen, M beta chain, D6S221E, DMB, major histocompatibility complex, class II, DM beta, MHC class II antigen DMB, MHC class II antigen HLA-DM beta chain, MHC class II HLA-DMB, Really interesting new gene 7 protein, RING7HLA class II histocompatibility antigen, DM beta chain
Gene Symbols HLA-DMB
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:AGGFVAHVESTCLLDDAGTPKDFTYCISFNKDLLTCWDPEENKMAPCEFGVLNSLANVLSQHLNQKDTLMQRLRNGLQNCATHTQPFWGSLTNRTRPPSVQ
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Asthma, Diabetes Research, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3109
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.