Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA DOA Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB095551 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB095551 100 μL
NB095552 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB095551 Supplier Novus Biologicals Supplier No. NBP303621100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HLA DOA Polyclonal specifically detects HLA DOA in Human samples. It is validated for Western Blot.

Specifications

Antigen HLA DOA
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias HLA class II histocompatibility antigen, DO alpha chain, HLA-D0-alpha, HLA-DNAHLADZ, HLA-DZA, lymphocyte antigen, major histocompatibility complex, class II, DN alpha, major histocompatibility complex, class II, DO alpha, MHC class II antigen DOA, MHC DN-alpha, MHC DZ alpha
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 26-135 of human HLA DOA (NP_002110.1).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Asthma, Diabetes Research, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3111
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.