Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA DOB Antibody (DOB 10E6/5), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP20420A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP20420A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP20420A94 Supplier Novus Biologicals Supplier No. NBP250420AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HLA DOB Monoclonal specifically detects HLA DOB in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen HLA DOB
Applications Western Blot, ELISA
Classification Monoclonal
Clone DOB 10E6/5
Conjugate Alexa Fluor 594
Dilution Western Blot, ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias DOB, FLJ57033, HLA class II histocompatibility antigen, DO beta chain, major histocompatibility complex, class II, DO beta, MHC class II antigen DOB
Gene Symbols HLA-DOB
Host Species Mouse
Immunogen C-terminal peptide MHC class II
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3112
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.