Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA DPA Antibody (TAL3C3), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP20601A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP20601A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP20601A32 Supplier Novus Biologicals Supplier No. NBP250601AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HLA DPA Monoclonal specifically detects HLA DPA in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen HLA DPA
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone TAL3C3
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias DP alpha 1 chain, HLA class II histocompatibility antigen, DP alpha chain, HLADP, HLA-SB alpha chain, major histocompatibility complex, class II, DP alpha 1, MHC class II antigen, MHC class II DP3-alpha, MHC class II DPA1, MHC class II HLA-DPA1 antigen, Primed lymphocyte test-1
Gene Symbols HLA-DPA1
Host Species Mouse
Immunogen Bristol 8 separated alpha chain preparation
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3113
Test Specificity TAL 3C3 is reactive on fixed tissue, reacts to class II antigens and cross-reacts with smooth muscle. Not cytotoxic with rabbit complement. TAL 3C3 detects alpha chain only by western blot and not the dimer.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.