Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA DPA Antibody (TAL3C3), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB309228 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309228 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309228 Supplier Novus Biologicals Supplier No. NBP250601MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HLA DPA Monoclonal antibody specifically detects HLA DPA in Human samples. It is validated for Western Blot, Immunohistochemistry, Radioimmune Assays

Specifications

Antigen HLA DPA
Applications Western Blot, Immunohistochemistry, Radioimmune Assays (RIA)
Classification Monoclonal
Clone TAL3C3
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Immunohistochemistry, Radioimmunoassay
Formulation 50mM Sodium Borate
Gene Alias DP alpha 1 chain, HLA class II histocompatibility antigen, DP alpha chain, HLADP, HLA-SB alpha chain, major histocompatibility complex, class II, DP alpha 1, MHC class II antigen, MHC class II DP3-alpha, MHC class II DPA1, MHC class II HLA-DPA1 antigen, Primed lymphocyte test-1
Host Species Mouse
Immunogen Bristol 8 separated alpha chain preparation
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Asthma, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3113
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.