Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA DPB1 Antibody (BRA-FB6), Novus Biologicals™
SDP

Catalog No. NBP24498601 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1 mg
0.2 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP24498601 0.1 mg
NBP24498600 0.02 mg
NBP24498602 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP24498601 Supplier Novus Biologicals Supplier No. NBP2449860.1MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence

HLA DPB1 Monoclonal specifically detects HLA DPB1 in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunofluorescence.

Specifications

Antigen HLA DPB1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Monoclonal
Clone BraFB6
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml
Formulation 10mM PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Alias DP beta 1 chain, DPB1, HLA class II histocompatibility antigen, DP(W4) beta chain, HLA DP14-beta chain, HLA-DP histocompatibility type, beta-1 subunit, HLA-DP1B, HLA-DPB, major histocompatibility complex class II HLA DPB1 protein, major histocompatibility complex, class II, DP beta 1, MHC class II antigen beta chain, MHC class II antigen DP beta 1 chain, MHC class II antigen DPB1, MHC class II antigen DPbeta1, MHC class II HLA-DP-beta, MHC HLA DPB1
Gene Symbols HLA-DPB1
Host Species Mouse
Immunogen Non-T, non-B human acute lymphoblastic leukemia REH6 cell line
Molecular Weight of Antigen 34 kDa
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Asthma, Diabetes Research, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3115
Test Specificity Recognizes a non-polymorphic determinant of DP-MHC class II. MHC class II antigens are transmembrane glycoproteins of non-covalently linked alpha (33-35kDa) and beta (27-30kDa) chains. It reportedly reacts with B- & non-T, non-B cell lines but not with T- and myeloid cell lines and leukemias. Differential expression of MHC class II antigens on fetal and adult lymphocytes, malignant B cells appears to reflect the stage of cell differentiation which may be useful in the study of lymphoproliferative disorders.
Target Species Human
Content And Storage Store at 4C.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.