Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA DQ Antibody (HLA-DQA1/2866R), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB378835 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB378835 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB378835 Supplier Novus Biologicals Supplier No. NBP308747AF488
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

HLA DQ Monoclonal antibody specifically detects HLA DQ in Human, Porcine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen)

Specifications

Antigen HLA DQ
Applications Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone HLA-DQA1/2866R
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias CD, CELIAC1DQ alpha 1 chain, DC-1 alpha chain, DQ-A1, FLJ27088, FLJ27328, GSE, HLA class II histocompatibility antigen, DQ(W3) alpha chain, HLA-DCA, HLA-DQA, leucocyte antigen DQA1, leukocyte antigen alpha chain, major histocompatibility complex, class II, DQ alpha 1, MGC149527, MHC class II antigen, MHC class II DQA1, MHC class II HLA-D alpha glycoprotein, MHC class II HLA-DQ-alpha-1, MHC class II surface glycoprotein, MHC HLA-DQ alpha
Host Species Rabbit
Immunogen Recombinant full-length human HLA DQ protein
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Asthma, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3117
Target Species Human, Pig
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.