Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA DQ Antibody (TAL 4.1), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB1002EAPC Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002EAPC 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002EAPC Supplier Novus Biologicals Supplier No. NB1002700APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HLA DQ Monoclonal specifically detects HLA DQ in Human samples. It is validated for CyTOF, ELISA, Flow Cytometry, Immunohistochemistry, Western Blot.

Specifications

Antigen HLA DQ
Applications CyTOF, ELISA, Flow Cytometry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone TAL 4.1
Conjugate APC
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias CD, CELIAC1DQ alpha 1 chain, DC-1 alpha chain, DQ-A1, FLJ27088, FLJ27328, GSE, HLA class II histocompatibility antigen, DQ(W3) alpha chain, HLA-DCA, HLA-DQA, leucocyte antigen DQA1, leukocyte antigen alpha chain, major histocompatibility complex, class II, DQ alpha 1, MGC149527, MHC class II antigen, MHC class II DQA1, MHC class II HLA-D alpha glycoprotein, MHC class II HLA-DQ-alpha-1, MHC class II surface glycoprotein, MHC HLA-DQ alpha
Gene Symbols HLA-DQA1
Host Species Mouse
Immunogen Whole cells from the B line WT46
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Asthma, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3117
Test Specificity Human Leukocyte Antigen DQ1 (includes DQ5 and 6)
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.