Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA DQ/DR/DP Antibody (Bu26), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. N1064162A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1064162A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1064162A32 Supplier Novus Biologicals Supplier No. NB10064162AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HLA DQ/DR/DP Monoclonal specifically detects HLA DQ/DR/DP in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen HLA DQ/DR/DP
Applications Flow Cytometry
Classification Monoclonal
Clone Bu26
Conjugate Alexa Fluor 532
Dilution Flow Cytometry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CD, CELIAC1DQ alpha 1 chain, DC-1 alpha chain, DQ-A1, FLJ27088, FLJ27328, GSE, HLA class II histocompatibility antigen, DQ(W3) alpha chain, HLA-DCA, HLA-DQA, leucocyte antigen DQA1, leukocyte antigen alpha chain, major histocompatibility complex, class II, DQ alpha 1, MGC149527, MHC class II antigen, MHC class II DQA1, MHC class II HLA-D alpha glycoprotein, MHC class II HLA-DQ-alpha-1, MHC class II surface glycoprotein, MHC HLA-DQ alpha
Gene Symbols HLA-DQA1
Host Species Mouse
Immunogen Made to Human HLA DP DQ DR
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3117
Test Specificity NB100-64162 recognizes the beta chains of class II human major histocompatibility (MHC) antigens. The antibody reacts with DP, DQ, DR beta chains, which are expressed by antigen presenting cells, B- cells, monocytes and activated T lymphocytes.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.