Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA DQ/DR/DP Antibody (WR18) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017567 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017567 0.025 mg
NB10064358 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017567 Supplier Novus Biologicals Supplier No. NB100643580.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HLA DQ/DR/DP Monoclonal specifically detects HLA DQ/DR/DP in Human samples. It is validated for Flow Cytometry, Functional Assay, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation.

Specifications

Antigen HLA DQ/DR/DP
Applications Flow Cytometry, Functional Assay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone WR18
Concentration 1.0 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry 1:50-1:200, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Functional
Formulation PBS with 0.09% Sodium Azide
Gene Alias CD, CELIAC1DQ alpha 1 chain, DC-1 alpha chain, DQ-A1, FLJ27088, FLJ27328, GSE, HLA class II histocompatibility antigen, DQ(W3) alpha chain, HLA-DCA, HLA-DQA, leucocyte antigen DQA1, leukocyte antigen alpha chain, major histocompatibility complex, class II, DQ alpha 1, MGC149527, MHC class II antigen, MHC class II DQA1, MHC class II HLA-D alpha glycoprotein, MHC class II HLA-DQ-alpha-1, MHC class II surface glycoprotein, MHC HLA-DQ alpha
Gene Symbols HLA-DQA1
Host Species Mouse
Immunogen Human HLA Class II (DP, DQ, DR)
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Asthma, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3117
Test Specificity Recognizes HLA Class II beta chains of human major histocompatability antigen, Mw 28 kD. It reacts with a monomorphic determinant common to each beta chain type. The antigen is expressed by human B lymphocytes, monocytes and activated T lymphocytes.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.