Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA DQ/DR/DP Antibody (WR18) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $499.50
Specifications
Antigen | HLA DQ/DR/DP |
---|---|
Clone | WR18 |
Dilution | Flow Cytometry 1:50-1:200, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Functional |
Applications | Immunohistochemistry (Paraffin), Flow Cytometry, Immunohistochemistry, Functional Assay |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017567
![]() |
Novus Biologicals
NB100643580.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NB10064358
![]() |
Novus Biologicals
NB10064358 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
HLA DQ/DR/DP Monoclonal specifically detects HLA DQ/DR/DP in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
HLA DQ/DR/DP | |
Flow Cytometry 1:50-1:200, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Functional | |
Monoclonal | |
Purified | |
RUO | |
Human | |
3117 | |
Human HLA Class II (DP, DQ, DR) | |
Primary | |
Recognizes HLA Class II beta chains of human major histocompatability antigen, Mw 28 kD. It reacts with a monomorphic determinant common to each beta chain type. The antigen is expressed by human B lymphocytes, monocytes and activated T lymphocytes. |
WR18 | |
Immunohistochemistry (Paraffin), Flow Cytometry, Immunohistochemistry, Functional Assay | |
Unconjugated | |
Mouse | |
Adaptive Immunity, Asthma, Immunology | |
CD, CELIAC1DQ alpha 1 chain, DC-1 alpha chain, DQ-A1, FLJ27088, FLJ27328, GSE, HLA class II histocompatibility antigen, DQ(W3) alpha chain, HLA-DCA, HLA-DQA, leucocyte antigen DQA1, leukocyte antigen alpha chain, major histocompatibility complex, class II, DQ alpha 1, MGC149527, MHC class II antigen, MHC class II DQA1, MHC class II HLA-D alpha glycoprotein, MHC class II HLA-DQ-alpha-1, MHC class II surface glycoprotein, MHC HLA-DQ alpha | |
HLA-DQA1 | |
IgG2a | |
Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title