Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA DQ1/DQ3 Antibody (HL-37), FITC, Novus Biologicals™
SDP

Catalog No. NB359321 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB359321 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB359321 Supplier Novus Biologicals Supplier No. NB500405F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HLA DQ1/DQ3 Monoclonal antibody specifically detects HLA DQ1/DQ3 in Human, Porcine samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, CyTOF

Specifications

Antigen HLA DQ1/DQ3
Applications Western Blot, Flow Cytometry, Immunoprecipitation, CyTOF
Classification Monoclonal
Clone HL-37
Conjugate FITC
Dilution Western Blot, Flow Cytometry, Immunoprecipitation, CyTOF-ready
Formulation PBS
Gene Alias CELIAC1, HLA class II histocompatibility antigen, DQ beta 1 chain, HLA-DQB, IDDM1, MHC class II antigen DQB1, MHC class II antigen HLA-DQ-beta-1, MHC class II DQ beta chain, MHC class II HLA-DQ beta glycoprotein
Host Species Mouse
Immunogen Burkitt's lymphoma cell line Raji
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Asthma, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3119
Target Species Human, Pig
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.