Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA DQ3 Antibody (2HB6), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. N102701A750 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N102701A750 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N102701A750 Supplier Novus Biologicals Supplier No. NB1002701AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HLA DQ3 Monoclonal specifically detects HLA DQ3 in Human samples. It is validated for CyTOF, ELISA, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Radioimmune Assays (RIA), Western Blot.

Specifications

Antigen HLA DQ3
Applications CyTOF, ELISA, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Clone 2HB6
Conjugate Alexa Fluor 750
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CD, CELIAC1DQ alpha 1 chain, DC-1 alpha chain, DQ-A1, FLJ27088, FLJ27328, GSE, HLA class II histocompatibility antigen, DQ(W3) alpha chain, HLA-DCA, HLA-DQA, leucocyte antigen DQA1, leukocyte antigen alpha chain, major histocompatibility complex, class II, DQ alpha 1, MGC149527, MHC class II antigen, MHC class II DQA1, MHC class II HLA-D alpha glycoprotein, MHC class II HLA-DQ-alpha-1, MHC class II surface glycoprotein, MHC HLA-DQ alpha
Gene Symbols HLA-DQA1
Host Species Mouse
Immunogen ATL mouse spleen cells and PRIESS (HLA-DR4 ) cells.
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3117
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.