Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA DQA2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP152094 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP152094 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP152094 Supplier Novus Biologicals Supplier No. NBP152094
Add to Cart
Add to Cart

Goat Polyclonal Antibody

HLA DQA2 Polyclonal specifically detects HLA DQA2 in Human samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen HLA DQA2
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_064440.1
Gene Alias DX alpha chain, DX-ALPHA, HLA class II histocompatibility antigen, DQ alpha 2 chain, HLA class II histocompatibility antigen, DQ(6) alpha chain, HLA-DQA1, HLA-DXAMHC class II DQA2, major histocompatibility complex, class II, DQ alpha 2
Gene Symbols HLA-DQA2
Host Species Goat
Immunogen Peptide with sequence C-QGLRSVGASRH corresponding to C-Terminus according to NP_064440.1, NP_002113.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3118
Test Specificity This antibody is expected to recognize: NP_064440.1 (HLA-DQA2; GeneID: 3118) and also the very similar NP_002113.2 (HLA-DQA1; GeneID:3117).
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.