Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA DQB1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB167108 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167108 20 μg
NB167107 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB167108 Supplier Novus Biologicals Supplier No. NBP31600620UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HLA DQB1 Polyclonal antibody specifically detects HLA DQB1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen HLA DQB1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 50% glycerol, pH7.3
Gene Alias DC-1 alpha chain, DC-alpha, HLA class II histocompatibility antigen, DQ alpha 1 chain-like, HLA-DCA, MHC class II DQA1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 33-127 of human HLA DQB1 (NP_002114.3).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Asthma, Diabetes Research, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3119
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.